3630
Glycylglycine, Free Base
Excellent buffer above pH 8.0.
Synonym(s):
Glycylglycine, Free Base
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
>98% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
pKa (20 °C)
8.4
shipped in
ambient
storage temp.
15-25°C
InChI
1S/C4H8N2O3.ClH/c5-1-3(7)6-2-4(8)9;/h1-2,5H2,(H,6,7)(H,8,9);1H
InChI key
YHBAZQDEMYQPJL-UHFFFAOYSA-N
General description
Excellent buffer above pH 8.0 (pKa = 8.4 at 20°C).
Excellent buffer above pH 8.0.
Application
- Polyetheretherketone/magnesium composite selectively coated with hydroxyapatite for enhanced in vitro bio-corrosion resistance and biocompatibility: Describes the use of Ca(EDTA) in preparing bioactive composite materials, emphasizing its role in improving biocompatibility and corrosion resistance (HD Jung, HS Park, MH Kang, SM Lee, HE Kim, Y Estrin, 2014).
- Artificially controlled degradable inorganic nanomaterial for cancer theranostics: Discusses the application of EDTA-Ca2+ complexes in creating novel inorganic nanomaterials for targeted drug delivery and therapy in cancer treatment (Y Liu, G Zhang, Q Guo, L Ma, Q Jia, L Liu, J Zhou, 2017).
Warning
Toxicity: Standard Handling (A)
Preparation Note
Heating to 45°C may be required to achieve complete dissolution.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service