Skip to Content
MilliporeSigma
All Photos(1)

Documents

361562

Sigma-Aldrich

GSK-3β Inhibitor XVIII

The GSK-3β Inhibitor XVIII, also referenced under CAS 1139875-74-3, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3β Inhibitor XVIII, 2-(Chloro-4-(4-thiophen-2-yl-pyrimidin-2-ylamino)-phenyl)-(4-methyl-pierazin-1-yl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN5OS
CAS Number:
Molecular Weight:
413.92
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thienylpyrimidine compound that is reported to act as a potent and selective GSK-3β inhibitor (IC50 = 64 nM), while exhibiting little or no activity against 58 and much reduced potency against 7 (>75% inhibition at 5 µM) other kinases. Shown to activate cellular Wnt pathway signaling (EC50 = 1.25 µM in a Super (8X) TOPFlash reporter assay) in HEK293 cultures and promote proliferation of murine and hamster β cell lines (EC50 = 1.143 µM using murine R7T1 cultures) as well as of primary rat β cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Wang, W., et al. 2009. Proc. Natl. Acad. Sci. USA106, 1427.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weidong Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(5), 1427-1432 (2009-01-24)
To identify small molecules that can induce beta-cell replication, a large chemical library was screened for proliferation of growth-arrested, reversibly immortalized mouse beta cells by using an automated high-throughput screening platform. A number of structurally diverse, active compounds were identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service