Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

356790

Millipore

Glyphosine

A plant growth regulator that also enhances anti-B:9-23 TCR-dependent T cell activation by modulating murine class II MHC I-Ag7-mediated B:9-23 presentation on APCs.

Synonym(s):

Glyphosine, Cp41845, N,N-bis(Phosphonomethyl)glycine, 2-(bis(Phosphonomethyl)amino)acetic acid, Polaris, Cp41845, Polaris, N,N-bis(Phosphonomethyl)glycine, 2-(bis(Phosphonomethyl)amino)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO8P2
CAS Number:
Molecular Weight:
263.08
MDL number:
UNSPSC Code:
12352200
form:
solid
Assay:
≥98% (NMR)

Quality Level

Assay

≥98% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C4H11NO8P2/c6-4(7)1-5(2-14(8,9)10)3-15(11,12)13/h1-3H2,(H,6,7)(H2,8,9,10)(H2,11,12,13)

InChI key

OXHDYFKENBXUEM-UHFFFAOYSA-N

General description

A glycine-phosphonic acid compound, negatively charged at neutral pH, with immunomodulatory and anti-diabetic properties. Shown to enhance CD4+ T cell response to B-chain of insulin (amino acids 9-23) even at 10 nM levels (EC50 = 70.6 nM). Occupies a pocket 9 of I-Ag7 (the class II molecules of the diabetes-prone NOD mice) and alters TCR/peptide-MHC affinity. Stimulates IL-10 secretion in splenocyes in non-obese diabetic (NOD) mice. Daily administration of glyphosine is shown to delay the onset of diabetes in NOD mice. Acts as a competitive inhibitor of phoshoenolpyruvate carboxylase in plants.
A plant growth regulator that also enhances anti-B:9-23 (aa 9 to 23 of insulin B chain) TCR-dependent T cell activation (EC50 = 70.6 nM; ECmax = 500 nM) by modulating murine class II MHC I-Ag7-mediated B:9-23 presentation on APCs, without affecting the activation of anti-chromogranin T cells by chromogranin- I-Ag7 complex. Reported to augment B:9-23 in stimulating IL-10 production from splenocytes of diabetic NOD mice and PBMCs of type 1 diabetic individuals with HLA-DQ8, and effectively prevent the onset of diabetes in NOD mice (80 mg/kg via daily i.p.). Computer structural docking analysis indicates pocket 9 of the I-Ag7 binding groove as the glyphosine targeting site.

Packaging

Packaged under inert gas

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Michels, A.W., et al. 2011. J. Immunol.187, 5921.
Stadinski, B.D., et al. 2010. Proc. Natl. Acad. Sci. USA107, 10978.
Podesta, F.E., et al. 1987. Plant Cell Physiol.28, 375.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service