Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

354206

Sigma-Aldrich

Anti-Glutathione S-Transferase A1-1 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-GST A1-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSTA1(2938)

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the A1-1 form (α-form) of glutathione S-transferase (GST).
Recognizes the α-form of GST. Does not react with µ- or ϖ-GSTs.
This Anti-Glutathione S-Transferase A1-1 Rabbit pAb is validated for use in Immunoblotting for the detection of Glutathione S-Transferase A1-1.

Immunogen

Human
recombinant human GST A1-1 (Cat. No. 354205)

Application

Immunoblotting (1:1000; see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

Serum diluted in PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Holmes, J., et al. 1990. Br. J. Cancer62, 209.
Specific for the α-form of GST. Does not react with µ- or ϖ-forms of GST. For immunoblotting a dilution of 1:1000 will yield a positive result with ≥0.2 µg of human A1-1 GST, using alkaline phosphatase-conjugated anti-rabbit IgG as a secondary antibody. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoliang Fang et al.
Journal of cell science, 127(Pt 8), 1631-1639 (2014-02-21)
Centrosome cohesion, mostly regarded as a proteinaceous linker between parental centrioles, ensures that the interphase centrosome(s) function as a single microtubule-organizing center. Impairment of centrosome cohesion leads to the splitting of centrosomes. Although the list of cohesion proteins is growing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service