Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345798

Millipore

Genapol® X-100, PROTEIN GRADE® Detergent, 10% Solution, Sterile-Filtered

These detergents have been purified for the isolation of membrane proteins.

Synonym(s):

Polyoxyethylene (10) Isotridecyl Ether

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161902
NACRES:
NA.21

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

88

color

clear colorless

CMC

150 μM

shipped in

ambient

storage temp.

2-8°C

General description

GENAPOL® Detergents are polyglycol ethers with the general formula R-CH2-O-(CH2CH2O)nH, where n = the number of ethylene oxides. The Calbiochem′s GENAPOLs have been purified for the isolation of membrane proteins (cloud point: 82°C in H2O; HLB number:13-14). They can be used in protocols that require UV monitoring of the solubilized proteins because of their low UV absorbance. Since several investigators have reported problems associated with peroxide and aldehyde impurities in polyoxyethylene detergents, these GENAPOL detergents have been additionally purified to reduce levels of contaminating peroxides, aldehydes, metals, and salts (aldehyde concentration: <200 µM; peroxide concentration: <200 µM; conductivity: <50 µmhos).
GENAPOL® Detergents are polyglycol ethers with the general formula R-CH2-O-(CH2CH2O)nH, where n = the number of ethylene oxides. These detergents have been purified for the isolation of membrane proteins (cloud point: 82°C in H2O; HLB number: 13-14). They can be used in protocols that require UV monitoring of the solubilized proteins because of their low UV absorbance. Since several investigators have reported problems associated with peroxide and aldehyde impurities in polyoxyethylene detergents, these GENAPOL Detergents have been additionally purified to reduce levels of contaminating peroxides, aldehydes, metals, and salts (aldehyde concentration: <200 µM; peroxide concentration: <200 µM; conductivity: <50 µmhos).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

10% aqueous solution.

Other Notes

Ashani, Y., and Catravas, G.N. 1980. Anal. Biochem.109, 55.
Lever, M. 1977. Anal. Biochem.83, 274.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Genapol is a registered trademark of Clariant GmbH
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service