Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344510

Sigma-Aldrich

FTase Inhibitor I

The FTase Inhibitor I controls the biological activity of FTase. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

FTase Inhibitor I, N-[2(S)-[2(R)-Amino-3-mercaptopropylamino]-3-methylbutyl]-Phe-Met-OH, B581

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38N4O3S2
Molecular Weight:
470.69
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable inhibitor of farnesyltransferase (FTase) that is about 37-fold more active against FTase (IC50 = 21 nM in vitro) than against geranylgeranyltransferase (GGTase; IC50 = 790 nM). Very resistant to proteolysis.
A potent, cell-permeable, selective, peptidomimetic inhibitor of farnesyltransferase (FTase) that is approximately 37-fold more active against FTase (IC50 = 21 nM in vitro) than against geranylgeranyltransferase (GGTase; IC50 = 790 nM). Very resistant to proteolysis.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FTase
Product does not compete with ATP.
Reversible: no
Target IC50: 21 nM against FTase in vitro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yamaguchi, M., et al. 2004. Stroke35, 1750.
Cox, A.D., et al. 1994. J. Biol. Chem.269, 19203.
Garcia, A.M., et al. 1993. J. Biol. Chem. 268, 18415.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen M Deveau et al.
Biochemical pharmacology, 183, 114349-114349 (2020-11-28)
Serotonin neurotransmission is largely governed by the regulation of the serotonin transporter (SERT). SERT is modulated in part by cholesterol, but the role of cholesterol and lipid signaling intermediates in regulating SERT are unknown. Serotonergic neurons were treated with statins
Yusuke Marikawa et al.
Molecular human reproduction, 27(4) (2021-03-08)
Early embryos are vulnerable to environmental insults, such as medications taken by the mother. Due to increasing prevalence of hypercholesterolemia, more women of childbearing potential are taking cholesterol-lowering medications called statins. Previously, we showed that inhibition of the mevalonate pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service