Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344281-M

Millipore

Forskolin, NKH477

A cell-permeable and water-soluble Forskolin derivative that is ~1.87-fold more potent than Forskolin in stimulating type V adenylate cyclase (AC) activity, while exhibiting similar potency as Forskolin toward type II & III AC.

Synonym(s):

Forskolin, NKH477, 6-(3-Dimethylaminopropionyl)forskolin, HCl, Colforsin Daropate, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO8 · xHCl
CAS Number:
Molecular Weight:
509.63 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C27H43NO8.ClH/c1-10-24(5)15-18(31)27(33)25(6)17(30)11-13-23(3,4)21(25)20(35-19(32)12-14-28(8)9)22(34-16(2)29)26(27,7)36-24;/h10,17,20-22,30,33H,1,11-15H2,2-9H3;1H/t17-,20-,21-,22-,24-,25-,26+,27-;/m0./s1

InChI key

VIRRLEDAYYYTOD-YHEOSNBFSA-N

General description

A cell-permeable and water-soluble Forskolin (Cat. No. 344270) derivative that is ~1.87-fold more potent than Forskolin in stimulating type V adenylate cyclase (AC) activity, while exhibiting similar potency as Forskolin toward type II & III AC (1.04- & 0.89-fold relative to Forskolin). NKH477 is shown to be orally available, blood-brain barrier permeant, and display reactivity toward a broad-spectrum of species, including human, canine, guinea pig, murine, procine, and rat.
A cell-permeable and water-soluble Forskolin (Cat. No. 344270) derivative that is ~1.87-fold more potent than Forskolin in stimulating type V adenylate cyclase (AC) activity, while exhibiting similar potency as Forskolin toward type II & III AC (1.04- & 0.89-fold relative to Forskolin). NKH477 is shown to be orally available, blood-brain barrier permeant, and display reactivity toward a broad-spectrum of species, including human, canine, guinea pig, murine, procine, and rat.

Warning

Toxicity: Highly Toxic (H)

Other Notes

Nakashima, S., et al. 2005. J. Heart Lung Transplant.24, 462.
Morinobu, S., et al. 1999. J. Neurochem.72, 2198.
Satake, K., et al. 1998. Br. J. Pharmacol.123, 753.
Toya, Y., et al. 1998. J. Mol. Cell Cardiol.30, 97.
Takeuchi, M., et al. 1995. Cardiovasc. Drug Rev.13, 339.
Hosono, M., et al. 1992. J. Cardiovasc. Pharmacol.19, 625.
Shafiq, J., et al. 1992. Circ. Res.71, 70.
Hosono, M., et al. 1990. Eur. J. Pharmacol.183, 2110.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service