Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344280

Sigma-Aldrich

Fostriecin, Sodium Salt, Streptomyces pulveraceous - CAS 87860-39-7 - Calbiochem

An antitumor antibiotic that acts as a potent protein phosphatase 2A (PP2A) inhibitor (IC₅₀ = 3.2 nM).

Synonym(s):

Fostriecin, Sodium Salt, Streptomyces pulveraceous - CAS 87860-39-7 - Calbiochem, FST, Phosphotrienin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26NaO9P
CAS Number:
Molecular Weight:
452.37
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

colorless

solubility

ethanol: 10 mg/mL
methanol: 10 mg/mL
water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

An antitumor antibiotic that acts as a potent protein phosphatase 2A (PP2A) inhibitor (IC50 = 3.2 nM). Inhibits protein phosphatase 1 (PP1) at much higher concentrations (IC50 = 131 µM). Does not affect the activity of protein phosphatase 2B (PP2B). Inhibits the catalytic activity of DNA topoisomerase II (topo II) and arrests cell growth at the G2/M phase of the cell cycle in a dose-dependent manner.
Antitumor antibiotic that acts as a potent protein phosphatase 2A (PP2A) inhibitor (IC50 = 3.2 nM). Also inhibits protein phosphatase 1 (PP1) at higher concentrations (IC50 = 131 µM), but has no effect on protein phosphatase 2B (PP2B). Inhibits the catalytic activity of DNA topoisomerase II (topo II) and arrests cell growth at the G2/M phase of the cell cycle in a dose-dependent manner.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PP2a
Product does not compete with ATP.
Reversible: no
Target IC50: 3.2 nM, 131 µM, against protein phosphatase 2A (PP2A), protein phosphatase 1 (PP1), respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Cheng, A., et al. 1998. Cancer Res. 58, 3611.
Walsh, A.H., et al. 1997. FEBS Lett. 416, 230.
Boritzki, T.J., et al. 1988. Biochem. Pharmacol. 37, 4063.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service