Skip to Content
MilliporeSigma
All Photos(1)

Documents

341329

Sigma-Aldrich

Fatostatin

A cell-permeable diarylthiazole compound that prevents the cellular activation of the SREBP-1/2 precursors to the active nuclear forms by blocking SREBPs-SCAP ER to Golgi translocation.

Synonym(s):

Fatostatin, 125B11, 2-Propyl-4-(4-(p-tolyl)thiazol-2-yl)pyridine, HBr, SREBP Processing Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2S · xHBr
CAS Number:
Molecular Weight:
294.41 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

pale yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H18N2S.BrH/c1-3-4-16-11-15(9-10-19-16)18-20-17(12-21-18)14-7-5-13(2)6-8-14;/h5-12H,3-4H2,1-2H3;1H

InChI key

RJCFNQZVFUMORB-UHFFFAOYSA-N

General description

A cell-permeable diarylthiazole compound that prevents the cellular activation of the SREBP-1/2 (Sterol Regulatory Element Binding Proteins 1/2) precursors to the active nuclear forms (>95% inhibition at 20 µM in CHO-K1 cells) by blocking SREBPs-SCAP (SREBP Cleavage-Activating Protein) ER to Golgi translocation with little effect against the transcription activity of the mature SREBPs. Binding studies employing recombinant SCAP truncation constructs reveal that Fatostatin targets SCAP between aa 448 to 767 in a reversible manner and is of no effect against SREBP-SCAP interaction. Fatostatin effectively suppresses SREBPs-dependent cellular functions in vitro, such as insulin-induced 3T3-L1 adipogenesis (complete inhibition at ≤20 µg/ml) and IGF1-dependent DU-145 proliferation (IC50 = 100 nM), and exhibits in vivo efficacy in lowering blood glucose level (by 70%) and body weight (by 12%) of ob/ob mice over a 28-day treatment period (30 mg/kg daily i.p.) without affecting animal food intake or apparent adverse effects.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kamisuki, S., et al. 2009. Chem. Biol.16, 882.
Choi, Y., et al. 2003. J. Biol. Chem.278, 7320.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Athina Maniscalchi et al.
Redox biology, 71, 103074-103074 (2024-02-18)
Brain iron accumulation constitutes a pathognomonic indicator in several neurodegenerative disorders. Metal accumulation associated with dopaminergic neuronal death has been documented in Parkinson's disease. Through the use of in vivo and in vitro models, we demonstrated that lipid dysregulation manifests

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service