Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341298

Sigma-Aldrich

FASTsilver Gel Staining Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$457.00

$457.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$457.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$457.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service