Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324905

Sigma-Aldrich

Epirubicin Hydrochloride - CAS 56390-09-1 - Calbiochem

A cell-permeable antitumor antibiotic.

Synonym(s):

Epirubicin Hydrochloride - CAS 56390-09-1 - Calbiochem, 4ʹ-Epidoxorubicin, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO11 · xHCl
CAS Number:
Molecular Weight:
543.52 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange-red

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10-,13-,15-,17-,22-,27-;/m0./s1

InChI key

MWWSFMDVAYGXBV-FGBSZODSSA-N

General description

A cell-permeable, antitumor antibiotic. A stereoisomer of doxorubicin that exhibits reduced cardiotoxicity. Its antitumor actions are mediated by targeting topoisomerase II.
This product has been discontinued.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Antitumor antibiotic
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstiution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Luck, H.J., et al. 1996. Semin. Oncol.23, 33.
Sandri, M.I., et al. 1996. Br. J. Cancer73, 1518.
Boddy, A.V., et al. 1995. Eur. J. Cancer31A, 69.
Pouna, P., et al. 1995. Cancer. Chemother. Pharmacol.35, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramchandra Vigay Amnekar et al.
World journal of gastroenterology, 26(6), 598-613 (2020-02-28)
The prognosis of gastric cancer continues to remain poor, and epigenetic drugs like histone deacetylase inhibitors (HDACi) have been envisaged as potential therapeutic agents. Nevertheless, clinical trials are facing issues with toxicity and efficacy against solid tumors, which may be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service