Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

324889

Sigma-Aldrich

eIF2-GTP-tRNAiMet ternary complex Inhibitor Negative Control, NCPdCPU

The eIF2-GTP-tRNAiMet ternary complex Inhibitor Negative Control, NCPdCPU controls the biological activity of eIF2-GTP-tRNAiMet ternary complex.

Synonym(s):

eIF2-GTP-tRNAiMet ternary complex Inhibitor Negative Control, NCPdCPU, 1-(2-chloro-5-nitrophenyl)-3-(3,4-dichlorophenyl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$88.40

$88.40


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
$88.40

About This Item

Empirical Formula (Hill Notation):
C13H8Cl3N3O3
CAS Number:
Molecular Weight:
360.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$88.40


Please contact Customer Service for Availability

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL, yellow

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=C(Cl)C=CC([N+]([O-])=O)=C1)NC2=CC=C(Cl)C(Cl)=C2

InChI

1S/C13H8Cl3N3O3/c14-9-3-1-7(5-11(9)16)17-13(20)18-12-6-8(19(21)22)2-4-10(12)15/h1-6H,(H2,17,18,20)

InChI key

LHXSZFFCIQIFEH-UHFFFAOYSA-N

General description

A negative control for eIF2-GTP-tRNAiMet ternary complex Inhibitor, BTdCPU.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, T., et al. 2011. Nat. Chem. Biol.7, 610.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service