Skip to Content
MilliporeSigma
All Photos(1)

Documents

324690

Millipore

EMPIGEN® BB Detergent, 30% Solution

Synonym(s):

n-Dodecyl-N,N-dimethylglycine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161902
NACRES:
NA.25

form

solution

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear pale yellow

CMC

1.6 - 2.1 mM

shipped in

ambient

storage temp.

10-30°C

General description

A zwitterionic detergent. Lauryldimethylbetaine is useful as a detergent for solubilization of membrane components and for preparation of viral agents. Also useful for the solubilization of keratins. Has a critical micellar concentration of 1.6-2.1 mM.
A zwitterionic detergent. Lauryldimethylbetaine is useful as a detergent for solubilization of membrane components and for preparation of viral antigens. Also useful for solubilization of keratins.

Warning

Toxicity: Irritant (B)

Physical form

30% ±1% aqueous solution.

Preparation Note

Can be further diluted with H₂O to the desired concentration.

Other Notes

Lowthert, L.A., et al. 1995. Biochem. Biophys. Res. Commun. 206, 370.Mukklis, F.A., et al. 1986. Vaccine4, 191.James, L.T., and Heckels, J.E. 1981. J. Immunol. Methods42, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
EMPIGEN is a registered trademark of Huntsman Corporation

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service