Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324515-M

Millipore

eEF-2 Kinase Inhibitor, NH125

The eEF-2 Kinase Inhibitor, NH125, also referenced under CAS 278603-08-0, controls the biological activity of eEF-2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

eEF-2 Kinase Inhibitor, NH125, 1-Benzyl-3-cetyl-2-methylimidazolium iodide, 1-Cetyl-3-benzyl-2-methylimidazolium iodide, CaM-Dependent Kinase III Inhibitor, NH125

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45IN2
CAS Number:
Molecular Weight:
524.56
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white to yellow

solubility

DMSO: 20 mg/mL
70% ethanol: 5 mg/mL

storage temp.

2-8°C

SMILES string

[I-].[n+]2(c([n](cc2)CCCCCCCCCCCCCCCC)C)Cc1ccccc1

InChI

1S/C27H45N2.HI/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-19-22-28-23-24-29(26(28)2)25-27-20-17-16-18-21-27;/h16-18,20-21,23-24H,3-15,19,22,25H2,1-2H3;1H/q+1;/p-1

InChI key

RVWOHCBHAGBLLT-UHFFFAOYSA-M

General description

A cell-permeable imidazolium compound that inhibits the activity of bacterial histidine kinases (EnvZ, PhoQ, BvgS, and EvgS) and eukaryotic eEF-2 (elongation factor 2) kinase/CaMKIII (IC50 = 60 nM), while exhibiting much lower potency against PKC, PKA, and CaMKII (IC50 = 7.5, 80, and >100 µM, respectively). NIH125 is a known P-gp substrate, which limits its in vivo efficacy as an anticancer and antimicrobial agent.

Biochem/physiol Actions

Primary Target
Bacterial histidine kinases (EnvZ, PhoQ, BvgS, and EvgS) and eukaryotic eEF-2 (elongation factor 2) kinase/CaMKIII
Target IC50: 60 nM against the activity of bacterial histidine kinases (EnvZ, PhoQ, BvgS, and EvgS) and eukaryotic eEF-2 (elongation factor 2) kinase/CaMKIII

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Arora, S., et al. 2004. Mol. Pharmacol.66, 460.
Arora, S., et al. 2003. Cancer Res.63, 6894.
Yamamoto, K., et al. 2000. Biosci. Biotechnol. Biochem.64, 919.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service