Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324396-M

Millipore

Anti-Dopamine D2S Receptor Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

species reactivity

rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DRD2(1813)

General description

Anti-Dopamine D2S Receptor, rabbit polyclonal, recognizes the ~48 kDa dopamine D2S receptor. Does not cross-react with other dopamine receptors. It is validated for use in ELISA, WB, and IHC.
Rabbit polyclonal antibody supplied as lyophilized, undiluted serum. Recognizes the ~48 kDa dopamine D2s receptor.
Recognizes the ~48 kDa dopamine D25 receptor. Does not cross-react with other dopamine receptors.

Immunogen

Human
a synthetic (PLKEAARRC) peptide corresponding to amino acids 239-246 of human D2s dopamine receptor

Application

ELISA (see comments)

Immunoblotting (1:800)

Immunohistochemistry (1:5000)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute the antibody with 100 µl PBS containing 10 mg/ml BSA. For further dilution of the antibody, use a similar buffer formulation. Following reconstitution, refrigerate (4°C).

Other Notes

Does not cross-react with other dopamine receptors. This antibody has also been reported to work for ELISA. For immunoblotting, a 1:800 dilution of the reconstituted serum detected a single band at ~48 kDa in an analysis using whole rat brain homogenates. For immunohistochemistry, a 1:500 dilution of the reconstituted serum demonstrated specific staining of rat brain sections including the plasma membrane of neurons in the cortex and caudate putamen. Staining was also observed along apical dendrites of layer 5 cortex cells. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service