Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3125-12

Sigma-Aldrich

Oil Red O

HARLECO® For staining of lipids

Synonym(s):

Oil Red O, Sudan Red 5B

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
408.5
Colour Index Number:
26125
MDL number:
UNSPSC Code:
41116130
EC Index Number:
215-295-3

form

powder

mp

182-186 °C

λmax

520-530 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

wet ice

InChI

1S/C26H24N4O/c1-16-9-10-17(2)22(13-16)27-28-23-14-19(4)24(15-18(23)3)29-30-26-21-8-6-5-7-20(21)11-12-25(26)31/h5-15,29H,1-4H3/b28-27?,30-26-

InChI key

HJAYEODIXUYVIC-LTFAAIOASA-N

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
For staining of lipids.
HARLECO® For staining of lipids

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service