Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3000-OP

Millipore

Calcium chloride dihydrate

OmniPur®

Synonym(s):

Calcium chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.28

Quality Level

form

fine powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

176 °C (dec.) (lit.)

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Molecular biology grade.

Specifications:
Assay (as dihydrate); 99.0-105.0%
DNase: None detected
Heavy metals (as Pb): ≤0.0005%
Iron (Fe): ≤0.001%
Protease: None detected
RNase: None detected
Sulfate (SO4): ≤0.01%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service