Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263275

Millipore

Desmosine

A novel quaternary pyridinium poly-amino acid present in elastin.

Synonym(s):

Desmosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39N5O8
Molecular Weight:
525.60
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (TLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A novel quaternary pyridinium poly-amino acid present in elastin. Suggested to be responsible for the cross-linking of elastin leading to the formation of insoluble fibers. Urinary excretion of desmosine is a useful indicator of elastin breakdown resulting from increased elastase activity.
A novel quaternary pyridinium poly-amino acid present in elastin. Suggested to be responsible for the cross-linking of elastin leading to the formation of insoluble fibers. Urinary excretion of desmosine is a useful indicator of elastin breakdown resulting from increased elastase activity. Absorption max: 268 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Cross-linking of elastin
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Akagawa, M., and Suyama, K. 2000. Connect. Tissue Res.41, 131.
Bode, D.C., et al. 2000. Pulm. Pharmacol. Therap.13, 175.
Viglio, S., et al. 2000. Eur. Respir. J.15, 1039.
Davis, N.R., and Anwar, R.A. 1970. J. Am. Chem. Soc.92, 3778.
Thomas, J., et al. 1963. Nature200, 651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service