Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2618

Millipore

Deoxyribonucleic Acid, Sodium Salt, Calf Thymus

Purified DNA from calf thymus. A260/280 nm: ≥ 1.8); S20w: 15.4-17.4; hyperchromicity: ≥27%.

Synonym(s):

Deoxyribonucleic Acid, Sodium Salt, Calf Thymus, DNA, Calf Thymus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352208
NACRES:
NA.51

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A260/280 nm: ≥ 1.8); hyperchromicity: ≥27%.
Purified DNA from calf thymus.
Purified DNA from calf thymus. An essential component of chromosomes. A260/280 nm: ≥ 1.8); hypechromicity: ≥27%.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service