Skip to Content
MilliporeSigma
All Photos(1)

Documents

260960

Sigma-Aldrich

DNA-PK Inhibitor

The DNA-PK Inhibitor, also referenced under CAS 20357-25-9, controls the biological activity of DNA-PK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

DNA-PK Inhibitor, DMNB, 4,5- Di methoxy-2- nitro benzaldehyde, DNA-Dependent Protein Kinase Inhibitor, DMNB, 4,5-Dimethoxy-2-nitrobenzaldehyde, DNA-Dependent Protein Kinase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO5
CAS Number:
Molecular Weight:
211.17
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.77

Quality Level

Assay

≥90% (GC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

bright yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C9H9NO5/c1-14-8-3-6(5-11)7(10(12)13)4-9(8)15-2/h3-5H,1-2H3

InChI key

YWSPWKXREVSQCA-UHFFFAOYSA-N

General description

A cell-permeable vanillin derivative that acts as a potent and selective inhibitor of DNA-PK (DNA-dependent protein kinase) activity (IC50 = 15 µM) and DNA-PK-mediated DSB (double strand break) DNA repair by NHEJ (non-homologous DNA-end-joining). Shown to effectively sensitize cells to killing by Cisplatin (Cat. No. 232120). Does not affect PKC or Chk2 kinase activities.
A cell-permeable vanillin derivative that acts as a potent and selective inhibitor of DNA-dependent protein kinase (DNA-PK) activity (IC50 = 15 µM) and DNA-PK-mediated double strand break (DSB) DNA repair by non-homologous DNA-end-joining (NHEJ). Reported to effectively sensitize cells to Cisplatin (Cat. No. 232120). Does not affect the activities of PKC or Chk2.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA-PK activity
Product does not compete with ATP.
Reversible: no
Target IC50: 15 µM against DNA-PK (DNA-dependent protein kinase) activity

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Willmore, E., et al. 2004. Blood,103, 4659.
Durant, S., and Karran, P. 2003. Nucleic Acids Res.31, 5501.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service