Skip to Content
MilliporeSigma
All Photos(1)

Documents

252750

Millipore

Deferoxamine Mesylate

Iron chelating agent

Synonym(s):

Deferoxamine Mesylate, Desferrioxamine Mesylate, DFOM, Iron Chelator II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48N6O8 · xCH4O3S
CAS Number:
Molecular Weight:
560.68 (free base basis)
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

faint yellow

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C25H48N6O8.CH4O3S/c1-21(32)29(37)18-9-3-6-16-27-22(33)12-14-25(36)31(39)20-10-4-7-17-28-23(34)11-13-24(35)30(38)19-8-2-5-15-26;1-5(2,3)4/h37-39H,2-20,26H2,1H3,(H,27,33)(H,28,34);1H3,(H,2,3,4)

InChI key

IDDIJAWJANBQLJ-UHFFFAOYSA-N

General description

Iron chelating agent. Completely protects against dopamine-induced cell death. Has also been shown to interfere with hydroxyl radical formation, which mediates tissue damage in several disease states. Shows anti-proliferative effect on vascular smooth muscle cells in vitro and in vivo. Can also inhibit the oxidative chemistry of peroxynitrite by reaction of the hydroxamic acid moieties with trans-peroxynitrous acid.

Application


  • Defective ferritinophagy and imbalanced iron metabolism in PBDE-47-triggered neuronal ferroptosis and salvage by Canolol.: This study explores the application of Deferoxamine Mesylate in mitigating iron overload and oxidative stress in neuronal cells, demonstrating its potential in neuroprotection and treatment strategies for neurodegenerative disorders (Wang et al., 2024).

  • Ferroptosis resists intracellular Vibrio splendidus AJ01 mediated by ferroportin in sea cucumber Apostichopus japonicus.: The study demonstrates the potential of Deferoxamine Mesylate in aquatic animal health, particularly in enhancing the resistance against bacterial infections by modulating ferroptosis pathways (Wang et al., 2024).

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Other Notes

Bergeron, R.J., et al. 1996. J. Med. Chem. 39, 1575.
Ben-Shachar, D., et al. 1995. J. Neurochem. 64, 718.
Denicola, A., et al. 1995. Free Radic. Biol. Med. 19, 11.
Dzwigaj, S., and pererat, H., 1995. Free Radic. Res.14, 103.
Dang, S., et al. 1994. Res. Commun. Mol. Pathol. Pharmacol. 86, 43.
Porreca, E., et al. 1994. Arterioscler. Thromb. 14, 299.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Divya Venkatesh et al.
Genes & development, 34(7-8), 526-543 (2020-02-23)
MDM2 and MDMX, negative regulators of the tumor suppressor p53, can work separately and as a heteromeric complex to restrain p53's functions. MDM2 also has pro-oncogenic roles in cells, tissues, and animals that are independent of p53. There is less

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service