Skip to Content
MilliporeSigma
All Photos(1)

Documents

251515-M

Sigma-Aldrich

DAF-FM

A photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~3 nM.

Synonym(s):

DAF-FM, 3-Amino, 4-aminomethyl-2ʹ,7ʹ-difluorofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14F2N2O5
CAS Number:
Molecular Weight:
412.34
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

fluorescence

λex ~500 nm
λem ~515 nm

storage temp.

−20°C

InChI

1S/C21H14F2N2O5/c1-25-13-3-2-8-18(19(13)24)20(28)30-21(8)9-4-11(22)14(26)6-16(9)29-17-7-15(27)12(23)5-10(17)21/h2-7,25-27H,24H2,1H3

InChI key

DIJCILWNOLHJCG-UHFFFAOYSA-N

General description

A photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~3 nM. Reacts with NO in the presence of O2, resulting in a triazolo-fluorescein analog (DAF-FM T) that exhibits about 160-fold greater fluorescence quantum efficiency. The fluorescence intensity is reported to be stable above pH 5.8.
A photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~3 nM. Reacts with NO, in the presence of oxygen, resulting in the formation of a triazolo-fluorescein analog (DAF-FM T) that exhibits about 160-fold greater fluorescence quantum efficiency. The fluorescence intensity is reported to be stable above pH 5.8. Excitation max.: ~500 nm; emission max.: ~515 nm.

Biochem/physiol Actions

Primary Target
Nitric oxide (NO) fluorescent indicator

Warning

Toxicity: Irritant (B)

Physical form

In DMSO.

Reconstitution

Following initial thaw, aliquot and freeze at -20°C. Use immediately after dilution, discard any unused portion.

Other Notes

Kojima, H., et al. 2001. J. Neurochem.76, 1404.
Park. J.M., et al. 2001. Br. J. Pharmacol.132, 1876.
Itoh, Y., et al. 2000. Anal. Biochem.287, 203.
Kojima, H., et al. 1999. Angew. Chem. Int. Ed.38, 3209.

Legal Information

Manufactured by Daiichi Pure Chemicals Co., Ltd
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service