Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

243005

Millipore

L-Cysteine, Free Base

Naturally-occurring non-essential amino acid.

Synonym(s):

L-Cysteine, Free Base, Cys

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$83.80
100 G
$324.00
1 KG
$904.00

$83.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$83.80
100 G
$324.00
1 KG
$904.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO2S
CAS Number:
Molecular Weight:
121.16
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.54

$83.80


Please contact Customer Service for Availability

Request a Bulk Order

description

Merck USA index - 14, 2781

Quality Level

Assay

≥98% (perchloric acid titration)

form

solid

optical activity

[α]20/D +8 to +9°, c = 5 in 1 M HCl

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 CFU/g Total microorganisms
≤24 EU/g Endotoxin

color

white

solubility

water: 10 mg/mL
1 M HCl: 50 mg/mL

cation traces

Fe: ≤10 ppm
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

15-25°C

SMILES string

SCC([N+H3])C(=O)[O-]

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)

InChI key

XUJNEKJLAYXESH-UHFFFAOYSA-N

General description

Naturally occurring non-essential amino acid.
Naturally-occurring non-essential amino acid.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Aqueous stock solutions are stable for up to 24 h at -20°C. Acidic stock solutions, when aliquoted and stored at -20°C, are stable for up to 1 month.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service