Skip to Content
MilliporeSigma
All Photos(1)

Documents

235427

Sigma-Aldrich

Caspase-3 Inhibitor I, Cell-Permeable

InSolution, ≥90%

Synonym(s):

InSolution Caspase-3 Inhibitor I, Cell-Permeable, Cell-permeable, DEVD-CHO; CPP32/Apopain Inhibitor, Cell-permeable

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H158N20O27
Molecular Weight:
2000.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable inhibitor of caspase-3. The C-terminal DEVD-CHO sequence of this peptide is a highly specific, potent, and reversible inhibitor of caspase-3 (Ki<1 nM) that has also been shown to strongly inhibit PARP cleavage in cultured human osteosarcoma cell extracts (IC50 = 200 pM). The N-terminal sequence (residues 1-16) corresponds to the hydrophobic region (h-region) of the signal peptide of Kaposi fibroblast growth factor (K-FGF) and confers cell-permeability to the peptide.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-3
Product does not compete with ATP.
Reversible: yes
Target IC50: 200 pM inhibiting PARP cleavage in cultured human osteosarcoma cell extracts

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Glu-Val-Asp-CHO

Physical form

A 5 mM (1 mg/100 µl) solution of Caspase-3 Inhibitor I, Cell-permeable (Cat. No. 235423) in DMSO.

Reconstitution

Following initial thaw, aliquot, purge with inert gas, and freeze (-20°C).

Other Notes

Lin, Y.-Z., et al. 1996. J. Biol. Chem. 271, 5305.
Nicholson, D.W. 1996. Nature Biotechnol. 14, 297.
Schlegel, J., et al. 1996. J. Biol. Chem.271, 1841.
Lin, Y.-Z., et al. 1995. J. Biol. Chem.270, 14255.
Nicholson, D.W., et al. 1995. Nature 376, 37.
Tewari, M., et al. 1995. Cell 81, 801.
Lazebnik, Y.A., et al. 1994. Nature371, 346.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service