Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

234567M

Sigma-Aldrich

Concanavalin A, Canavalia ensiformis

Concanavalin A, Canavalia ensiformis, CAS 11028-71-0, is a carbohydrate-binding protein that agglutinates various animal cells. Restores the growth pattern of tumor cells to that of normal cells.

Synonym(s):

Concanavalin A, Canavalia ensiformis, Con A, Jack Bean Hemagglutinin

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$307.00
1 G
$1,010.00

$307.00


Available to ship onMarch 31, 2025Details



Select a Size

Change View
250 MG
$307.00
1 G
$1,010.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.42

$307.00


Available to ship onMarch 31, 2025Details


Quality Level

description

Merck USA index - 14, 2495

form

lyophilized

potency

≤20 μg/mL lectin agglutination activity (as determined from serial dilutions of a 1 mg/mL solution)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

pI 

4.5 - 5.5

storage temp.

2-8°C

General description

Carbohydrate-binding protein that agglutinates various animal cells. Also known to restore the growth pattern of tumor cells to that of normal cells. T lymphocyte activator. Has binding sites for metal ions, α-D-mannose, and α-D-glucose. Has a pI of 4.5-5.5.

Warning

Toxicity: Harmful (C)

Preparation Note

To dissolve, add Con A slowly to a solution of 10 mM HEPES buffered saline, 100 µM CaCl₂, pH 8.5. Allow the lectin to dissolve without agitation.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) or add 0.08% NaN₃ and refrigerate (4°C). Stock solutions are stable for up to 6 months at -20°C or for up to 3 months at 4°C.

Analysis Note

The agglutination activity is the lowest conc. to agglutinate a 2% suspension of human erythrocytes after 1 hr incubation at 25°C; requires type A human blood.

Other Notes

Gantner, F., et al. 1995. Hepatology 21, 190.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service