Skip to Content
MilliporeSigma
All Photos(1)

Documents

234184-M

Sigma-Aldrich

Collagen, Type II, Bovine Joint Cartilage

Synonym(s):

Collagen, Type II, Bovine Joint Cartilage

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized solid (Salt-free)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

0.5 M acetic acid: 1 mg/mL

storage temp.

−20°C

General description

Highly purified preparation isolated from the triple helical domain of guanidine-HCl-extracted and pepsin-digested bovine joint cartilage collagen Type II. Subunit composition: α1(II)3.
Highly purified preparation isolated from the triple helical domain of guanidine-HCl-extracted and pepsin-digested bovine joint cartilage collagen Type II. Subunit composition: α1(II)3. Does not react with anti-collagen Type I and anti-collagen Type III by Western blotting.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Solutions gradually solidify at room temperature.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Sieper, J., et al. 1996. Arthritis Rheum. 39, 41.
Boissier, M.-C., et al. 1990. Arthritis Rheum. 33, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service