Skip to Content
MilliporeSigma
All Photos(2)

Documents

23-048-M

Sigma-Aldrich

Parkin Protein, active, 10 µg

Active, full length, human recombinant human Parkin. N-terminal 6His-tagged. For use in Enzyme Assays. Functions as an E3 ligase in ubiquitination assays.

Synonym(s):

Parkinson juvenile disease protein 2, PARK2, PRKN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

100
300

recombinant

expressed in Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Parkin is an E3-ubiquitin ligase belonging to the RBR (RING – In Between RING – RING) family. Mutations in Parkin that result in a loss-of-function are the most frequent cause of familial Parkinson’s disease and are responsible for a large percentage of autosomal recessive juvenile parkinsonism (AR-JP). In addition to its auto-ubiquitination, Parkin can also ubiquitinate various substrates, including the CDK2-interacting protein cyclin E1, the a-synuclein-interacting protein synphilin-1, the synaptic vesicle-associated CDC-rel1 and the p38 subunit of aminoacyl-tRNA synthetase/JTV-1. Ubiquitination of substrates occurs in conjunction with the E2 enzymes UbcH7, UbcH8 or UbcH13/Uev1.

Application

Ubiquitination Cascade Comoponent: E3

Physical form

Purified using immobilized metal affinity chromatography.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service