Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

23-030-M

Sigma-Aldrich

SCF-Fbw7 Protein Complex, Active, 10 µg

Active complex of four (4) full length, recombinant human enzymes: Fbw7, Skp1, Cul1 & Rbx1. For use in Enzyme Assays. Functions as an E3 ligase in ubiquitination assays.

Synonym(s):

SCF Fbxw7, SCF hCdc4

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$831.00
250 μG
$4,920.00

$831.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
$831.00
250 μG
$4,920.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

$831.00


Please contact Customer Service for Availability

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

Gene Information

General description

N-terminal c-Myc, 6His-tagged, recombinant human Smad7 full length, expressed by baculovirus in Sf21 insect cells.
The SCF (Skp1-Cul1-F-box protein) complexes represent the largest family of ubiquitin-protein ligases and mediate the ubiquitination of a broad spectrum of regulatory and signalling proteins in diverse cellular pathways. The SCF consists of three invariant components, Skp1, Cul1, and Rbx1 and an interchangeable subunit, an F-box protein which is responsible for recruiting specific substrates to be ubiquitinated by the SCF. SCFFbw7 mediates the ubiquitination and subsequent proteasomal degradation of target proteins involved in cell division, growth and differentiation, including Cyclin E1, c-Myc, Notch and c-Jun. Mutations have been found in the Fbw7 subunit of SCFFbw7 in various types of human cancers, therefore Fbw7 is considered to be a potent tumour suppressor.

Application

Ubiquitination Cascade Comoponent: E3

Physical form

Purified using glutathione sepharose.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service