Skip to Content
MilliporeSigma
All Photos(2)

Documents

220491

Sigma-Aldrich

Chk2 Inhibitor II

InSolution, ≥95%

Synonym(s):

InSolution Chk2 Inhibitor II, 2-(4-(4-Chlorophenoxy)phenyl)-1H-benzimidazole-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14ClN3O2
CAS Number:
Molecular Weight:
363.80
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−20°C

InChI

1S/C20H14ClN3O2/c21-14-4-8-16(9-5-14)26-15-6-1-12(2-7-15)20-23-17-10-3-13(19(22)25)11-18(17)24-20/h1-11H,(H2,22,25)(H,23,24)

InChI key

UXGJAOIJSROTTN-UHFFFAOYSA-N

General description

A cell-permeable and reversible benzimidazolo compound that acts as a potent and ATP-competitive inhibitor of Chk2 with an IC50 of 15 nM and a Ki of 37 nM. Displays ~ 1,000-fold greater selectivity over Cdk1/B and CK1 (IC50 = 12 M and 17 M, respectively) and only weakly affects the activities of a panel of 31 kinases (<25% inhibition at 10 M), including Chk1. Shown to rescue both peripheral CD4+ and CD8+ T-cells from γ-irradiation-induced apoptosis with an EC50 of 3 M and 7.6 M, respectively. The solid form of this compound (Cat. No. 220486) is also available.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

Supplied as a 25 mM (5 mg/550 µL) solution of Chk2 Inhibitor II (Cat. No. 220486) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Arienti, K. L., et al. 2005. J. Med. Chem.48, 1873.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting-Yu Chen et al.
Cell death and differentiation, 28(6), 1865-1879 (2021-01-20)
The DNA-PK maintains cell survival when DNA damage occurs. In addition, aberrant activation of the DNA-PK induces centrosome amplification, suggesting additional roles for this kinase. Here, we showed that the DNA-PK-p53 cascade induced primary cilia formation (ciliogenesis), thus maintaining the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service