Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219672

Sigma-Aldrich

CFTR Inhibitor III, OXO-172

The CFTR Inhibitor III, OXO-172 controls the biological activity of CFTR. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

CFTR Inhibitor III, OXO-172, 3-((3-Trifluoromethyl)phenyl)-5-((4-carboxyphenyl)methylene)-4-thiazolidinone, OXO-172

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10F3NO4S
Molecular Weight:
393.34
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC, NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 1 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable CFTRinh-172 (Cat. No. 219670) 2-oxo analog that exhibits ~25-fold higher aqueous solubility than the 2-thioxo CFTRinh-172 (420 µM vs. 17 µM, respectively, in PBS with 2% DMSO at 25 °C) and great potency, albeit slightly weaker than that of CFTRinh-172, in inhibiting CFTR-mediated apical membrane chloride current upon 20 µM Forskolin (Cat. No. 344270) stimulation in CFTR-expressing FRT cells (IC50 = 1.4 µM vs. 0.38 µM with CFTRinh-172) with practically no cytotoxicity (91% of no-drug control FRT in viability assay; 48 h at 50 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sonawane, N.D., and Verkman, A.S. 2008. Bioorg. Med. Chem.16, 8187.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel J Henning et al.
Nature chemical biology, 18(4), 412-421 (2022-02-26)
Many diseases are driven by proteins that are aberrantly ubiquitinated and degraded. These diseases would be therapeutically benefited by targeted protein stabilization (TPS). Here we present deubiquitinase-targeting chimeras (DUBTACs), heterobifunctional small molecules consisting of a deubiquitinase recruiter linked to a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service