Skip to Content
MilliporeSigma
All Photos(1)

Documents

219506

Sigma-Aldrich

CBFβ-Runx1 Inhibitor II

Synonym(s):

RUNX1/CBFβ inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Application

The CBFΒ-Runx1 Inhibitor II, Ro5-3335, also referenced under CAS 30195-30-3, suppresses CBFΑ/Runx1-CBFΒ transactivation activity (25 µM; 293-0 cells) via direct bindings to both subunits of the heterodimeric transcription factor complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service