Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218829

Sigma-Aldrich

Caspase-3 Substrate IX, Fluorogenic

A non-fluorescent peptidyl (DEVD)-Rhodamine 110-bisamide that acts as a highly sensitive, photostable caspase-3 fluorogenic substrate.

Synonym(s):

Caspase-3 Substrate IX, Fluorogenic, (Z-DEVD)₂-Rh 110

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H78N10O27
Molecular Weight:
1515.44
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow

solubility

DMSO: 1 mg/mL

fluorescence

λex ~485 nm
λem ~535 nm

shipped in

wet ice

storage temp.

−20°C

General description

A non-fluorescent peptidyl (DEVD)-Rhodamine 110-bisamide that acts as a highly sensitive, photo-stable caspase-3 fluorogenic substrate. Reported to detect less than 1 ng/ml of caspase-3. Also reported to detect capase-3 activity in intact cells by flow cytometry. Useful for high throughput screening applications. Excitation max: ~485 nm; emission max: ~535 nm.
A non-fluorescent peptidyl (DEVD)-Rhodamine 110-bisamide that acts as a highly sensitive, photostable caspase-3 fluorogenic substrate. Reported to detect less than 1 ng/ml of caspase-3. Useful for high throughput screening applications.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A highly sensitive, photostable caspase-3 fluorogenic substrate
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

(Z-Asp-Glu-Val-Asp)₂-Rhodamine 110

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hug, H., et al. 1999. Biochemistry38, 13906.
Liu, J., et al. 1999. Bioorg. Med. Chem. Lett.9, 3231.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service