Skip to Content
MilliporeSigma
All Photos(1)

Documents

218805

Sigma-Aldrich

Caspase-9 Substrate II, Colorimetric

The Caspase-9 Substrate II, Colorimetric serves as the substrate for Caspase-9.

Synonym(s):

Caspase-9 Substrate II, Colorimetric, Ac-LEHD- pNA, Ac-LEHD-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38N8O11
Molecular Weight:
674.66
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A colorimetric substrate for caspase-9. Cleavage is monitored colorimetrically at ~405 nm.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Caspase-9
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Leu-Glu-His-Asp-pNA

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Thornberry, N.A., et al. 1997. J. Biol. Chem. 272, 17907.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service