Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217692

Sigma-Aldrich

CDC25 Phosphatase Inhibitor II, NSC 663284

The CDC25 Phosphatase Inhibitor II, NSC 663284, also referenced under CAS 383907-43-5, controls the biological activity of CDC25 Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

CDC25 Phosphatase Inhibitor II, NSC 663284, 6-Chloro-7-(2-morpholin-4-yl-ethylamino)quinoline-5,8-dione, DA3003-1, PTP Inhibitor XXI, VHR Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$279.00

$279.00


Please contact Customer Service for Availability


Select a Size

Change View
5 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C15H16ClN3O3
CAS Number:
Molecular Weight:
321.76
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$279.00


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 5 mg/mL (Use only fresh, anhydrous DMSO.)
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC1=C(C(=O)c3ncccc3C1=O)NCCN2CCOCC2

InChI

1S/C15H16ClN3O3/c16-11-13(18-4-5-19-6-8-22-9-7-19)15(21)12-10(14(11)20)2-1-3-17-12/h1-3,18H,4-9H2

InChI key

BMKPVDQDJQWBPD-UHFFFAOYSA-N

General description

A cell-permable, potent, irreversible, and competitive inhibitor of the Cdc25 phosphatase family (Ki = 29 nM, 95 nM, and 89 nM for Cdc25A, Cdc25B2, and Cdc25C, respectively) with anti-proliferative properties. Displays nearly 20- and 450-fold greater selectivity over VHR and PTP1B (IC50 = 4.0 µM and >100 µM, respectively). Reported to arrest cell cycle progression, inhibit Cdk dephosphorylation, and delay tumor growth (mean IC50 = 1.5 µM in NCI-60 tumor cell panel). Also shown to block ERK dephosphorylation caused by ectopic Cdc25A expression.
A cell-permeable 7-substituted quinolinedione compound that displays antiproliferative properties. Acts as a potent, irreversible and mixed competitive inhibitor of the CDC25 phosphatase family (Ki = 29 nM, 95 nM and 89 nM for CDC25A, CDC25B2 and CDC25C, respectively; IC50 = 210 nM for CDC25B2) and displays nearly 20- and 450-fold greater selectivity for the CDC252 phosphatases over VHR and PTP1B (IC50 = 4.0 µM and >100 µM, respectively). Reported to arrest cell cycle progression, inhibit Cdk dephosphorylation and delay tumor growth (IC50 = ~1.5 µM in NCI-60 tumor cell panel). Shown to modify CDC25A catalytic domain and to block cellular ERK dephosphorylation in HeLa-CDC25A cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CD25A
Product does not compete with ATP.
Reversible: no
Target Ki: 29 nM, 95 nM and 89 nM for CDC25A, CDC25B2 and CDC25C, respectively; 210 nM for CDC25B2

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 1 month at -20°C. Ethanol stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chao, J. I., et al. 2004. J. Biol. Chem.279, 20267.
Han, Y., et al. 2004. J. Pharmacol. Exp. Ther.309, 64.
Pu, L., et al. 2002. J. Biol. Chem.277, 46877.
Lazo, J.S., et al. 2001. J. Med. Chem.44, 4042.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

470.1 °F

Flash Point(C)

243.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service