Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217657

Sigma-Aldrich

Anti-CD49f Mouse mAb (BQ16)

liquid, >95% (SDS-PAGE), clone BQ16

Synonym(s):

Anti-VLA-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

BQ16, monoclonal

Assay

>95% (SDS-PAGE)

form

liquid

contains

0.5 mg/mL gentamycin sulfate as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-CD49f, mouse monoclonal, clone BQ16, recognizes the ~150 kDa CD49f (α6 integrin) in Raji cells. It is validated for FC, WB, IP, and IHC on frozen sections.
Protein A purified mouse monoclonal antibody. Recognizes the ~150 kDa CD49f (VLA-6) protein.
Recognizes the ~150 kDa CD49f (α6 integrin) in Raji cells.

Immunogen

UM-UC-9 cells (human bladder cancer cell line)

Application

Flow Cytometry (10 µg/ml)

Frozen Sections (see comments)

Immunoblotting (see comments)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Berditchevski, F., et al. 1995. J. Biol. Chem. 270, 17784.
Liebert, M., et al. 1993. Hybridoma 12, 67.
This antibody has also been reported to work for immunoblotting, immunoprecipitation, and frozen sections. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service