Skip to Content
MilliporeSigma
All Photos(2)

Documents

215901

Sigma-Aldrich

Carbonic Anhydrase IX/XII Inhibitor II, U-104

The Carbonic Anhydrase IX/XII Inhibitr II, U-104 controls the biological activity of Carbonic Anhydrase IX/XII. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Carbonic Anhydrase IX/XII Inhibitor II, U-104, CAIX Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12FN3O3S
Molecular Weight:
309.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell permeable ureido-sulfonamide Carbonic Anhydrase (CA) inhibitor that is selective for human hCA IX and hCA XII (Ki = 45 nM and 4.5 nM, respectively) against two other physiologically relevant hCA isoforms (Ki = 5080 nM and 9640 nM for hCA I and hCA II, respectively). It elicits anti-tumor growth effects dose-dependently from 19 to 38 mg/kg in a mouse tumor growth model implanted with MDA-MB-231 LM2-4Luc+ cells, and demonstrates metastasis inhibition in a mouse metastasis model injected with 4T1 cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Lock, F.E., et al. 2012. Oncogenein press.
Pacchiano, F., et al. 2011. J Med Chem.54, 1896.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Han et al.
Scientific reports, 11(1), 8626-8626 (2021-04-23)
Liver cancer is the fatal cause of cancer deaths worldwide due to its aggressiveness and lack of effective therapies. Tiliroside (C30H26O13) is an active compound extracted from herb plant Tribulus terrestris L., which has been used as alternative therapy in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service