Skip to Content
MilliporeSigma
All Photos(1)

Documents

208690

Sigma-Aldrich

Calmodulin, Bovine Brain

Calmodulin is a ubiquitous Ca2+-binding protein that serves as a physiological effector of a wide range of biological processes. Purified from bovine brain.

Synonym(s):

Calmodulin, Bovine Brain, CaM, Ca 2+/Calmodulin, CaM, Ca2+/Calmodulin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

specific activity

≥12,500 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

A ubiquitous Ca2+-binding protein distributed in eukaryotic cells that serves as a major physiological effector for a wide range of cellular responses. The Ca2+-calmodulin complex is involved in the maintenance of intracellular Ca2+ homeostasis, as well as in the regulation of the intracellular concentration of other second messengers, such as cyclic AMP and inositol trisphosphate. Calmodulin purified from bovine brain.



Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.
A ubiquitous Ca2+-binding protein that serves as a physiological effector of a wide range of biological processes. Calmodulin purified from bovine brain.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of calmodulin that gives rise to 50% of the maximal enzyme activation of a standard level of activator-deficient 3ʹ,5ʹ-cyclic nucleotide phosphodiesterase.

Physical form

Lyophilized from 150 mM NaCl, 50 mM Tris-HCl, 2 mM EDTA, pH 7.6.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C or -70°C). Stock solutions are stable for up to 2 months at -20°C or for up to 4 months at -70°C.

Other Notes

Carl, A., et al. 1990. J. Physiol.429, 205.
Appleman, M.M., et al. 1985. J. Cyclic Nucleotide and Protein Phosph. Res.10, 417.
Cheung, W.T. 1979. Science207, 19.
Sharma, R.K., and Wang, J.H. 1979. Adv. Cyclic Nucleotide Res.10, 187.
Wang, J.H., and Waisman, D.M. 1979. Curr. Topics Cell. Reg.15, 47.
Watterson, A., et al. 1976. J. Biol. Chem.251, 4501.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service