Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2080-OP

Millipore

Agarose

OmniPur® Grade, Super-Fine Resolution

Synonym(s):

OmniPur® Agarose Super-Fine Resolution, 3,6-Anhydro-α-L-galacto-β-D-galactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105317

Quality Level

form

fine powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Suitable for microsatellite analysis, RFLP and AFLP and analytical and preparative electrophoresis. Provides 2% resolution in the 200 bp-1,000 bp range.

Specifications:
Ash: ≤0.35%
Conductivity (1%, water): ≤10 µmho
DNase: None detected
EEO (-mr): ≤0.12
Gelling range (3%): ≤35.5°C
Gel strength (1.5%): ≥500 g/cm
Melting point (1.5%): ≤70.0°C Moisture (H2O) ≤7.0%
Protease: None detected
RNase: None detected
Sulfate (SO4): ≤0.11%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service