Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203987

Sigma-Aldrich

Butein

A cell-permeable, reversible, substrate competitive, plant polyphenol that acts as a specific protein tyrosine kinase inhibitor.

Synonym(s):

Butein, 2ʹ,4ʹ,3,4-Tetrahydroxychalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C15H12O5/c16-10-3-4-11(14(19)8-10)12(17)5-1-9-2-6-13(18)15(20)7-9/h1-8,16,18-20H/b5-1+

InChI key

AYMYWHCQALZEGT-ORCRQEGFSA-N

General description

A cell-permeable, reversible, substrate competitive, plant polyphenol that acts as a specific protein tyrosine kinase inhibitor. Potently inhibits the tyrosine kinase activity of the EGF receptor (IC50 = 65 µM) and p60c-src (IC50 = 65 µM) The inhibition is competitive with respect to ATP and non-competitive with respect to the substrate. Butein is also a potent antioxidant and anti-inflammatory agent. Has been shown to inhibit glutathione reductase (GSH-RD) and rat liver glutathione S-transferase (GST; IC50 = 9 µM). Reported to activate sirtuins and promote the survival of eukaryotic cells.
A cell-permeable, reversible, substrate competitive, plant polyphenol that acts as a specific protein tyrosine kinase inhibitor. Potently inhibits the tyrosine kinase activity of the EGF receptor (IC50 = 65 µM) and p60c-src (IC50 = 65 µM). The inhibition is competitive with respect to ATP and non-competitive with respect to the substrate. Butein is also a potent antioxidant and anti-inflammatory agent. Has been shown to inhibit glutathione reductase and rat liver glutathione-S-transferase (IC50 = 9 µM). Reported to activate sirtuins and promote the survival of eukaryotic cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Tyrosine kinase activity of the EGF receptor
Product competes with ATP.
Reversible: yes
Target IC50: 65 µM against tyrosine kinase activity of the EGF receptor

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 weeks at -20°C.

Other Notes

Howitz, K.T., et al. 2003. Nature425, 191.
Chan, S.C., et al. 1998. Planta Med.64, 153.
Yang, E.B., et al. 1998. Biochem. Biophys. Res. Commun. 245, 435.
Zhang, K., et al. 1997. Biochem. Pharmacol. 54, 1047.
Zhang, K., and Das, N.P. 1994. Biochem. Pharmacol. 47, 2063.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service