Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203702

Sigma-Aldrich

Brain Derived Neurotrophic Factor, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human brain drived neurotrophic factor expressed in E. coli. Promotes survival and growth of neurons in the central nervous system. Sequence analysis indicates that brain derived neurotrophic factor is structurally and functionally related to nerve growth factor. mRNA for brain derived neurotrophic factor is found predominantly in the central nervous system, whereas mRNA for nerve growth factor is also found in many non-CNS tissues.
Recombinant, human brain drived neurotrophic factor expressed in E. coli. Promotes survival and growth of neurons. Structurally and functionally related to nerve growth factor. Its actions are mediated via the TrkB receptor.

Biochem/physiol Actions

Supports survival of rat basal forebrain primary septal cultured neurons. Induces choline acetyltransferase (EC₅₀ = 50 ng/ml)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions.
Reconstitute to a concentration of ≥10 µg/ml with sterile distilled H₂O or aqueous buffers. For more dilute solutions, add a stabilizing protein such as 0.1% BSA or HSA.

Other Notes

Erickson, J.T., et al. 1996. J. Neurosci. 16, 5361.
Leibrock, J., et al. 1989. Nature341, 149.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Mooney et al.
Neuroscience, 179, 256-266 (2011-02-01)
A second wave of neuronal generation occurs in the ventrobasal nucleus of the rat thalamus (VB) during the first three postnatal weeks. The present study tested the hypotheses (1) that postnatal neurogenesis in the VB is neurotrophin-regulated and (2) that
Christophe Heinrich et al.
Nature protocols, 6(2), 214-228 (2011-02-05)
Instructing glial cells to generate neurons may prove to be a strategy to replace neurons that have degenerated. Here, we describe a robust protocol for the efficient in vitro conversion of postnatal astroglia from the mouse cerebral cortex into functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service