Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203401

Millipore

Bleomycin Sulfate, Streptomyces verticillus

Mixture of cytotoxic glycopeptides that inhibits DNA synthesis by reacting with DNA and causing strand scission.

Synonym(s):

Bleomycin Sulfate, Streptomyces verticillus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (A₂ + B₂, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C55H83N17O21S3.H2O4S/c1-20-33(69-46(72-44(20)58)25(12-31(57)76)64-13-24(56)45(59)82)50(86)71-35(41(26-14-61-19-65-26)91-54-43(39(80)37(78)29(15-73)90-54)92-53-40(81)42(93-55(60)88)38(79)30(16-74)89-53)51(87)66-22(3)36(77)21(2)47(83)70-34(23(4)75)49(85)63-10-8-32-67-28(18-94-32)52-68-27(17-95-52)48(84)62-9-7-11-96(5)6;1-5(2,3)4/h14,17-19,21-25,29-30,34-43,53-54,64,73-75,77-81H,7-13,15-16,56H2,1-6H3,(H13-,57,58,59,60,61,62,63,65,66,69,70,71,72,76,82,83,84,85,86,87,88);(H2,1,2,3,4)/t21?,22?,23?,24?,25?,29-,30+,34?,35?,36?,37+,38+,39-,40-,41?,42-,43-,53+,54-;/m0./s1

InChI key

WUIABRMSWOKTOF-OCBSMOPSSA-N

General description

A mixture of cytotoxic glycopeptides that inhibits DNA synthesis by reacting with DNA and causing strand scission. Acts by generating hydroxyl free radicals which cleave N-glycosidic bonds and cause base elimination (mainly T) and phosphodiester bond scission. An antineoplastic agent capable of mimicking apoptosis. Useful in the treatment of squamous cell carcinomas, testicular tumors, and malignant lymphomas.
Mixture of cytotoxic glycopeptides that inhibits DNA synthesis by reacting with DNA and causing strand scission. Acts by generating hydroxyl free radicals which cleave N-glycosidic bonds and cause base elimination (mainly T) and phosphodiester bond scission. An anti-neoplastic agent. Bleomycin has been used in the treatment of squamous cell carcinomas, testicular tumors, and malignant lymphomas.

Biochem/physiol Actions

Potency: 1.5-2.0 U/mg

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short-term storage. Aqueous stock solutions (≥3 units/ml) are stable for up to 1 month at 4°C or for up to 3 months at -20°C.

Other Notes

Tounekki, O., et al. 1994. Bull. Cancer 81, 1043.
Chauvergne, J., et al. 1993. Bull. Cancer 80, 70.
Ikeda, S., et al. 1991. Anal. Biochem.192, 96.
Hait, W.N., et al. 1990. Cancer Res.50, 6636.
Arkin, H., et al. 1989. Cancer Res.49, 6556.
Wu, J.C., et al. 1983. J. Biol. Chem.258, 4694.
Takita, T., et al. 1968. Antibiotics21, 79.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phuong-Uyen C Dinh et al.
Nature communications, 11(1), 1064-1064 (2020-03-01)
Idiopathic pulmonary fibrosis (IPF) is a fatal and incurable form of interstitial lung disease in which persistent injury results in scar tissue formation. As fibrosis thickens, the lung tissue loses the ability to facilitate gas exchange and provide cells with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service