Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203112

Sigma-Aldrich

Biotin-NHS

Synonym(s):

Biotin-NHS, Biotin N-hydroxysuccinimide Ester

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H19N3O5S
CAS Number:
Molecular Weight:
341.38
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.41

Assay

≥90% (NMR)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S1C(C3NC(=O)NC3C1)CCCCC(=O)ON2C(=O)CCC2=O

InChI

1S/C14H19N3O5S/c18-10-5-6-11(19)17(10)22-12(20)4-2-1-3-9-13-8(7-23-9)15-14(21)16-13/h8-9,13H,1-7H2,(H2,15,16,21)

InChI key

YMXHPSHLTSZXKH-UHFFFAOYSA-N

General description

Biotinylates amino acids, peptides, or proteins under mild conditions.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, Aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinichiro Okamoto et al.
STAR protocols, 2(1), 100230-100230 (2020-12-29)
We developed an adeno-associated virus (AAV) vector-based technique to label mouse neostriatal neurons comprising direct and indirect pathways with different fluorescent proteins and analyze their axonal projections. The AAV vector expresses GFP or RFP in the presence or absence of
Denis Bedoret et al.
The Journal of clinical investigation, 119(12), 3723-3738 (2009-11-13)
The respiratory tract is continuously exposed to both innocuous airborne antigens and immunostimulatory molecules of microbial origin, such as LPS. At low concentrations, airborne LPS can induce a lung DC-driven Th2 cell response to harmless inhaled antigens, thereby promoting allergic
Chen Jiang et al.
Nature communications, 12(1), 1308-1308 (2021-02-28)
The precise spatiotemporal control of cell proliferation is key to the morphogenesis of epithelial tissues. Epithelial cell divisions lead to tissue crowding and local changes in force distribution, which in turn suppress the rate of cell divisions. However, the molecular
Dominic A Helmerich et al.
ACS nano, 14(10), 12629-12641 (2020-08-18)
The degree of labeling (DOL) of antibodies has so far been optimized for high brightness and specific and efficient binding. The influence of the DOL on the blinking performance of antibodies used in direct stochastic optical reconstruction microscopy (dSTORM) has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service