Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20-113

Sigma-Aldrich

Magnesium/ATP Cocktail

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$186.00

$186.00


Please contact Customer Service for Availability


Select a Size

Change View
1 ML
$186.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

$186.00


Please contact Customer Service for Availability

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

For use in kinase assays.
Research Category
All

Quality

Routinely evaluated by Kinase Assay.

Physical form

20mM MOPS, pH 7.2, 25mM β-glycerophosphate, 5mM EGTA, 1mM Na3VO4, 1mM dithiothreitol, 75mM MgCl2, and 0.5mM ATP

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The kinases aurora B and mTOR regulate the G1-S cell cycle progression of T lymphocytes.
Jianxun Song,Shahram Salek-Ardakani,Takanori So,Michael Croft
Nature Immunology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service