Skip to Content
MilliporeSigma
All Photos(1)

Documents

20-111

Sigma-Aldrich

Assay Dilution Buffer II (ADBII)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

For use in kinase assays.
Research Category
All

Physical form

20mM MOPS, pH 7.2, 25mM β-glycerol phosphate, 1mM sodium orthovanadate, 1mM dithiothreitol, 1mM CaCl2

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of carbohydrate recognition domains of pertussis toxin in adherence of Bordetella pertussis to human macrophages.
J van't Wout,W N Burnette,V L Mar,E Rozdzinski,S D Wright,E I Tuomanen
Infection and Immunity null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service