Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200980-M

Millipore

Bikinin

A cell-permeable nonsteroidal pyridinylamido compound that acts as an ATP-competitive inhibitor.

Synonym(s):

Bikinin, Arabidopsis GSK-3 Inhibitor, Arabidopsis SHAGGY-related Kinase Inhibitor, GSK-3-like Kinase BIN2 Inhibitor, 4-((5-Bromo-2-pyridinyl)amino)-4-oxobutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9BrN2O3
CAS Number:
Molecular Weight:
273.08
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

Brc1cnc(cc1)NC(=O)CCC(=O)O

InChI

1S/C9H9BrN2O3/c10-6-1-2-7(11-5-6)12-8(13)3-4-9(14)15/h1-2,5H,3-4H2,(H,14,15)(H,11,12,13)

InChI key

XFYYQDHEDOXWGA-UHFFFAOYSA-N

General description

A cell-permeable nonsteroidal pyridinylamido compound that acts as an ATP-competitive inhibitor against 7 of the 10 Arabidopsis thaliana GSK3′s, BIN2 & ASKα/ γ/ε/ζ/ι/θ, but not ASKβ/δ or three other Arabidopsis Ser/Thr kinases, AtMPK4, AtMPK6, and AtAURORA1. The inhibitory activity is plant species-selective, as Bikinin exhibits much reduced activity against human Gsk-3β, Aurora B, Aurora C (45.6%, 69.4%, and 42.8% inhibition, respectively, with 10 µM Bikinin), and little or no activity against 74 other human kinases, nor does it affect GSK-3β-mediated dorsoventral axis formation during Xenopus embryo development. Bikinin at 30 µM is shown to bypass brassinolide (BL) and BRI1 and effectively activate brassinosteroid (BR) signaling responses in Arabidopsis thaliana seedings, including decreased BES1 phosphorylation, increased hypocotyl length, long and bending petioles, blade-shaped, pale-green leaves, as well as reduced lateral root density.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Rybel, B.D., et al. 2009. Chem. Biol.16, 594.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service