Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

200275

Sigma-Aldrich

Berberine Chloride

A highly potent and selective oxysterol EBI2 (GPR183) agonist (Kd) = 450 pM in a saturation binding assay, and EC₅₀ = 140 pM over EC₅₀ = 2.1 nM for its enantiomer, 7β,25-OHC, in a GTP-γS binding assay).

Synonym(s):

Berberine chloride form, Berberine, Natural Yellow 18

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNO4
CAS Number:
Molecular Weight:
371.81
Colour Index Number:
75160
Beilstein:
3836585
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (NMR if no UV signal, HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

water: 1 mg/mL
DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

COC1=C(C=[N+](CCC2=C3C=C4C(OCO4)=C2)C3=C5)C5=CC=C1OC.[Cl-]

InChI

1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1

InChI key

VKJGBAJNNALVAV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

A highly potent and selective oxysterol EBI2 (GPR183) agonist (Kd) = 450 pM in a saturation binding assay, and EC50 = 140 pM over EC50 = 2.1 nM for its enantiomer, 7β,25-OHC, in a GTP-γS binding assay). Dose-dependently suppresses forskolin-induced cAMP accumulation in an EBI2-expressing SK-N-MC/CRE-β-galactosidase cell line (IC50 = 2 nM), but not in control cells. Stimulates migration of LPS-activated spleen B-cells and anti-CD3/CD28-activated CD4+ T-cells in a dose-dependent manner. In addition, pharmacological inhibition of its biosynthesis in vivo by Clotrimazole, a CYP7B1inhibitor, promotes the migration of adoptively transferred pre-activated B cells to the T/B boundary, mimicking the phenotype of pre-activated B cells in EBI2-deficient mice.

Application

Fluorescent stain for heparin in mast cells

Biochem/physiol Actions

An alkaloid with weak antibiotic properties. Substrate for MDR efflux pumps. Antimicrobial activities of berberine is potentiated by the MDR inhibitor 5´-methoxyhydnocarpin (5´-MHC). Berberine upregulates the expression of Pgp in hepatoma cells. Treatment with berberine potentially results in the reduced accumulation of chemotherapeutic drugs.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kinm. B., et al. 2011, Brit. Journ. Pharm.164, 106.
Kong, W., et al. 2004, Nature Med.10, 1344.
Dimlich, R. et al. 1980, Stain Technol.55, 217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingying Kou et al.
Frontiers in pharmacology, 11, 616251-616251 (2020-12-29)
Gastric cancer is one of the most common malignancies ranks as the second leading cause of cancer-related mortality in the world. Cisplatin (DDP) is commonly used for gastric cancer treatment, whereas recurrence and metastasis are common because of intrinsic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service