196810
Bax-Inhibiting Peptide, V5
A cell-permeable pentapeptide based on the Ku70-Bax inhibiting domain that offers cytoprotection.
Synonym(s):
Bax-Inhibiting Peptide, V5, BIP-V5, H-VPMLK-OH
Select a Size
$165.00
Select a Size
About This Item
$165.00
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
white
solubility
water: 1 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
Biochem/physiol Actions
Bax mediated apoptosis
Packaging
Warning
Sequence
Reconstitution
Other Notes
Sawada, M., et al. 2003. Nature Cell Biol.5, 352.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service