Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

196806

Sigma-Aldrich

Bax Channel Blocker II, iMAC2 - CAS 335166-00-2 - Calbiochem

A MAC (Mitochondrial Apoptosis-induced Channel) inhibitor iMAC1 tructural analog that is reported to be less potent than iMAC1 in blocking Bid-induced cytochrome c release in cell-free mitochondria preparations.

Synonym(s):

Bax Channel Blocker II, iMAC2 - CAS 335166-00-2 - Calbiochem, (±)-3,6-Dibromo-9-(2-fluoro-3-piperazin-1-yl-propyl)-carbazole, 2HCl, MAC Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20Br2FN3 · 2HCl
CAS Number:
Molecular Weight:
542.11
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A MAC (Mitochondrial Apoptosis-induced Channel) inhibitor iMAC1 (Cat. No. 196805) structural analog that is reported to be less potent than iMAC1 in blocking Bid-induced cytochrome c release in cell-free mitochondria preparations (40% vs. 77% inhibition with 5 µM respective inhibitor), but more effective than iMAC1 in inhibiting STS- (Cat. No. 569396 & 569397) induced cytochrome c release in intact FL5.12 cells (Complete vs ~35% inhibition with 5 µM respective inhibitor), presumably due to an enhanced permeability. iMAC2 is also shown to be a superior blocker of MAC-mediated current flow in reconstituted mitochondrial outer membrane patches from apoptotic FL5.12 cells (IC50 = 28 vs 300 nM in the case of iMAC2 and iMAC1, respectively) and protect FL5.12 from STS-induced apoptosis (IC50 = 2.5 µM).
A MAC (Mitochondrial Apoptosis-induced Channel) inhibitor iMAC1 (Cat. No. 196805) structural analog that is reported to be less potent than iMAC1 in blocking Bid-induced cytochrome c release in cell-free mitochondria preparations (40% vs. 77% inhibition with 5 µM respective inhibitor), but more effective than iMAC1 in inhibiting STS- (Cat. No. 569396 & 569397) induced cytochrome c release in intact FL5.12 cells (Complete vs ~35% inhibition with 5 µM respective inhibitor), presumably due to an enhanced permeability. iMAC2 is also shown to be a superior blocker of MAC-mediated current flow in reconstituted mitochondrial outer membrane patches from apoptotic FL5.12 cells (IC50 = 28 vs 300 nM in the case of iMAC2 and iMAC1, respectively) and protect FL5.12 from STS-induced apoptosis (IC50 = 2.5 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Peixoto, P.M., et al. 2009. Biochem. J.423, 381.
Bombrun, A., et al. 2003. J. Med. Chem.46, 4365.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service