Skip to Content
MilliporeSigma
All Photos(1)

Documents

196805

Sigma-Aldrich

Bax Channel Blocker

A cell-permeable dibromocarbazolo-piperazinyl derivative that displays anti-apoptotic properties.

Synonym(s):

Bax Channel Blocker, (±)-1-(3,6-Dibromocarbazol-9-yl)-3-piperazin-1-yl-propan-2-ol, iMAC1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21Br2N3O
CAS Number:
Molecular Weight:
467.20
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable dibromocarbazole-piperazinyl derivative that displays anti-apoptotic properties. Reported to effectively block Bid-induced cytochrome c release from HeLa cell mitochondria (~80% inhibition at 5 µM) by inhibiting Bax channel activity.
A cell-permeable dibromocarbazolo-piperazinyl derivative that displays anti-apoptotic properties. Effectively blocks Bid-induced cyctochrome c release from HeLa cell mitochondria (~80% inhibition at 5 µM) by inhibiting Bax channel-forming activity (IC50 = 520 nM in a liposome channel assay).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Bax-channel forming activity
Product does not compete with ATP.
Reversible: no
Target IC50: 520 nM blocks Bid-induced cyctochrome c release from HeLa cell mitochondria by inhibiting Bax channel-forming activity in a liposome channel assay

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Peixoto, P., et al. 2009. Biochem J.423, 381.
Bombrun, A., et al. 2003. J. Med. Chem.46, 4365.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dulguun Amgalan et al.
Nature cancer, 1(3), 315-328 (2020-08-11)
Doxorubicin remains an essential component of many cancer regimens, but its use is limited by lethal cardiomyopathy, which has been difficult to target, owing to pleiotropic mechanisms leading to apoptotic and necrotic cardiac cell death. Here we show that BAX

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service