Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93801

Supelco

Zinc oxide

EMPLURA®

Synonym(s):

Zinc oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
OZn
CAS Number:
Molecular Weight:
81.39
MDL number:
UNSPSC Code:
12352303

product line

EMPLURA®

Quality Level

Assay

98.5%

form

solid

manufacturer/tradename

EMPLURA®

mp

>1,000  °C ((> 1,832 °F) at ca. 1,013.25 hPa)

storage temp.

room temp

SMILES string

O=[Zn]

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service